Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103845.3 | NP_001097315.1 | 3 - 4 | 917 | 72 | |
NM_001202000.2 | NP_001188929.1 | 5 - 6 | 1297 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |