Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: norpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014720.2 | NP_001014720.1 | 12 - 13 | 3953 | 63 | |
NM_001014721.3 | NP_001014721.1 | 11 - 12 | 3491 | 63 | ||
NM_001169190.2 | NP_001162661.1 | 13 - 14 | 4222 | 63 | ||
NM_001297931.1 | NP_001284860.1 | 12 - 13 | 3616 | 63 | ||
NM_080330.4 | NP_525069.2 | 11 - 12 | 3491 | 63 | ||
NM_167008.2 | NP_726925.1 | 12 - 13 | 3953 | 63 | ||
GenBank mRNA | AF181641.1 | - | 11 - 12 | 3474 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |