Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103427.3 | NP_001096897.1 | 11 - 12 | 3751 | 72 | |
NM_001258625.2 | NP_001245554.1 | 12 - 13 | 3784 | 72 | ||
NM_132124.5 | NP_572352.1 | 10 - 11 | 3247 | 72 | ||
NM_167091.3 | NP_727125.1 | 10 - 11 | 3208 | 72 | ||
NM_167092.3 | NP_727126.1 | 10 - 11 | 3207 | 72 | ||
NM_167093.2 | NP_727127.1 | 10 - 11 | 3147 | 72 | ||
NM_167094.2 | NP_727128.1 | 10 - 11 | 3216 | 72 | ||
GenBank mRNA | BT003254.1 | - | 10 - 11 | 3194 | 72 | |
BT124988.1 | - | 12 - 13 | 3764 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |