Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103427.3 | NP_001096897.1 | 7 - 8 | 2246 | 70 | |
NM_001258625.2 | NP_001245554.1 | 7 - 8 | 2246 | 70 | ||
NM_132124.5 | NP_572352.1 | 6 - 7 | 1742 | 70 | ||
NM_167091.3 | NP_727125.1 | 6 - 7 | 1703 | 70 | ||
NM_167092.3 | NP_727126.1 | 6 - 7 | 1702 | 70 | ||
NM_167093.2 | NP_727127.1 | 6 - 7 | 1642 | 70 | ||
NM_167094.2 | NP_727128.1 | 6 - 7 | 1711 | 70 | ||
GenBank mRNA | BT003254.1 | - | 6 - 7 | 1689 | 70 | |
BT124988.1 | - | 7 - 8 | 2226 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |