Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103427.3 | NP_001096897.1 | 10 - 11 | 3413 | 65 | |
NM_001258625.2 | NP_001245554.1 | 11 - 12 | 3446 | 65 | ||
NM_132124.5 | NP_572352.1 | 9 - 10 | 2909 | 65 | ||
NM_167091.3 | NP_727125.1 | 9 - 10 | 2870 | 65 | ||
NM_167092.3 | NP_727126.1 | 9 - 10 | 2869 | 65 | ||
NM_167093.2 | NP_727127.1 | 9 - 10 | 2809 | 65 | ||
NM_167094.2 | NP_727128.1 | 9 - 10 | 2878 | 65 | ||
GenBank mRNA | BT003254.1 | - | 9 - 10 | 2856 | 65 | |
BT124988.1 | - | 11 - 12 | 3426 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |