Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272478.1 | NP_001259407.1 | 3 - 4 | 1289 | 64 | |
NM_001369992.1 | NP_001356939.1 | 3 - 4 | 1289 | 64 | ||
NM_079999.3 | NP_524738.1 | 3 - 4 | 1289 | 64 | ||
NM_167229.2 | NP_727418.1 | 4 - 5 | 1682 | 64 | ||
GenBank mRNA | AY069047.1 | - | 4 - 5 | 1865 | 64 | |
BT133196.1 | - | 3 - 4 | 1268 | 64 | ||
X56439.1 | - | 798 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |