Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132466.3 | NP_572694.1 | 5 - 6 | 1264 | 97 | |
NM_167274.2 | NP_727501.1 | 5 - 6 | 1264 | 97 | ||
NM_167276.2 | NP_727503.1 | 4 - 5 | 920 | 97 | ||
GenBank mRNA | AY058302.1 | - | 5 - 6 | 1247 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |