Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103480.2 | NP_001096950.2 | 4 - 5 | 822 | 91 | |
NM_001103481.3 | NP_001096951.3 | 3 - 4 | 653 | 91 | ||
NM_001144714.1 | NP_001138186.1 | 2 - 3 | 313 | 91 | ||
NM_001272516.2 | NP_001259445.2 | 3 - 4 | 653 | 91 | ||
NM_167278.3 | NP_727512.3 | 3 - 4 | 653 | 91 | ||
GenBank mRNA | BT010070.1 | - | 3 - 4 | 620 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |