Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rho-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080345.3 | NP_525084.1 | 1 - 2 | 485 | 62 | |
NM_167290.3 | NP_727538.1 | 1 - 2 | 485 | 62 | ||
GenBank mRNA | AF318285.1 | - | 1 - 2 | 459 | 62 | |
AY061605.1 | - | 2 - 3 | 789 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |