Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258706.2 | NP_001245635.1 | 6 - 7 | 3002 | 96 | |
NM_132540.3 | NP_572768.1 | 6 - 7 | 3002 | 96 | ||
NM_167313.3 | NP_727583.1 | 7 - 8 | 3037 | 96 | ||
GenBank mRNA | AY862156.1 | - | 6 - 7 | 2978 | 96 | |
BT010304.1 | - | 6 - 7 | 2975 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |