Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomosyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169264.2 | NP_001162735.1 | 22 - 23 | 4796 | 70 | |
NM_001169265.2 | NP_001162736.2 | 22 - 23 | 4547 | 70 | ||
NM_001258715.2 | NP_001245644.1 | 22 - 23 | 4148 | 70 | ||
NM_132591.5 | NP_572819.4 | 23 - 24 | 4193 | 70 | ||
NM_167328.4 | NP_727628.3 | 23 - 24 | 4836 | 70 | ||
NM_167329.3 | NP_727629.2 | 22 - 23 | 4996 | 70 | ||
NM_167330.3 | NP_727630.3 | 22 - 23 | 4669 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |