Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomosyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169264.2 | NP_001162735.1 | 5 - 6 | 1177 | 115 | |
NM_001169265.2 | NP_001162736.2 | 5 - 6 | 1177 | 115 | ||
NM_001258715.2 | NP_001245644.1 | 5 - 6 | 1177 | 115 | ||
NM_132591.5 | NP_572819.4 | 6 - 7 | 1222 | 115 | ||
NM_167328.4 | NP_727628.3 | 6 - 7 | 1217 | 115 | ||
NM_167329.3 | NP_727629.2 | 5 - 6 | 1377 | 115 | ||
NM_167330.3 | NP_727630.3 | 5 - 6 | 1299 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |