Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258496.3 | NP_001245425.1 | 13 - 14 | 9478 | 83 | |
NM_079884.5 | NP_524623.2 | 13 - 14 | 9484 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |