Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: axo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259701.1 | NP_001246630.1 | 6 - 7 | 1723 | 74 | |
NM_001259702.1 | NP_001246631.1 | 6 - 7 | 1723 | 74 | ||
NM_001259703.1 | NP_001246632.1 | 6 - 7 | 1723 | 74 | ||
NM_001274493.1 | NP_001261422.1 | 6 - 7 | 1832 | 74 | ||
NM_001274494.1 | NP_001261423.1 | 6 - 7 | 1832 | 74 | ||
NM_001274495.1 | NP_001261424.1 | 6 - 7 | 1832 | 74 | ||
NM_079917.5 | NP_524656.5 | 6 - 7 | 1832 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |