Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079918.3 | NP_524657.2 | 1 | 853 | 112 | |
GenBank mRNA | AY084113.1 | - | 1 | 817 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |