Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298883.1 | NP_001285812.1 | 4 - 5 | 1196 | 65 | |
NM_079928.2 | NP_524667.1 | 4 - 5 | 709 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster Inventoried |