Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169343.2 | NP_001162814.1 | 1 - 2 | 1789 | 121 | |
NM_001169344.2 | NP_001162815.1 | 3 - 4 | 1515 | 121 | ||
NM_001169345.2 | NP_001162816.1 | 2 - 3 | 432 | 121 | ||
NM_001169346.2 | NP_001162817.1 | 3 - 4 | 1443 | 121 | ||
NM_079929.3 | NP_524668.2 | 2 - 3 | 525 | 121 | ||
NM_167772.2 | NP_728449.1 | 2 - 3 | 570 | 121 | ||
NM_167773.3 | NP_728450.1 | 3 - 4 | 1519 | 121 | ||
NM_167774.3 | NP_728451.2 | 3 - 4 | 1474 | 121 | ||
GenBank mRNA | AF175711.2 | - | 2 - 3 | 553 | 121 | |
AF175713.1 | - | 416 | 121 | |||
AY121664.1 | - | 3 - 4 | 1525 | 121 | ||
BT021433.1 | - | 3 - 4 | 1518 | 121 | ||
BT099548.1 | - | 2 - 3 | 548 | 121 | ||
BT125869.1 | - | 2 - 3 | 489 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |