Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ect | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274748.1 | NP_001261677.1 | 6 - 7 | 1632 | 72 | |
NM_001300090.1 | NP_001287019.1 | 7 - 8 | 1755 | 72 | ||
NM_079967.6 | NP_524706.2 | 7 - 8 | 1788 | 72 | ||
NM_168400.3 | NP_729596.1 | 7 - 8 | 1731 | 72 | ||
GenBank mRNA | AY061556.1 | - | 7 - 8 | 1772 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |