Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sr-CII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201979.2 | NP_001188908.1 | 2 - 3 | 641 | 68 | |
NM_079981.3 | NP_524720.2 | 1 - 2 | 315 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |