Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298707.1 | NP_001285636.1 | 2 - 3 | 2473 | 96 | |
NM_079986.4 | NP_524725.2 | 2 - 3 | 2473 | 96 | ||
GenBank mRNA | AF145670.1 | - | 2 - 3 | 2450 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |