Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272159.1 | NP_001259088.1 | 4 - 5 | 4088 | 54 | |
NM_130478.3 | NP_569834.1 | 4 - 5 | 4088 | 54 | ||
GenBank mRNA | AY061125.1 | - | 4 - 5 | 4059 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |