Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272159.1 | NP_001259088.1 | 8 - 9 | 5657 | 73 | |
NM_130478.3 | NP_569834.1 | 9 - 10 | 5597 | 73 | ||
GenBank mRNA | AY061125.1 | - | 9 - 10 | 5568 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |