Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp52F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137249.4 | NP_611093.2 | 16 - 17 | 3762 | 91 | |
GenBank mRNA | BT133387.1 | - | 16 - 17 | 3742 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |