Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103866.2 | NP_001097336.1 | 13 - 14 | 4245 | 73 | |
NM_001299556.1 | NP_001286485.1 | 12 - 13 | 4175 | 73 | ||
NM_001299557.1 | NP_001286486.1 | 13 - 14 | 6125 | 73 | ||
NM_137272.2 | NP_611116.2 | 13 - 14 | 4452 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |