Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP54D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274131.2 | NP_001261060.1 | 3 - 4 | 1518 | 74 | |
NM_137392.3 | NP_611236.2 | 3 - 4 | 1714 | 74 | ||
GenBank mRNA | BT133065.1 | - | 3 - 4 | 1502 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |