Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdgBbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137404.5 | NP_611248.3 | 1 - 2 | 161 | 64 | |
GenBank mRNA | AF160934.1 | - | 1 - 2 | 127 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |