Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nopo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137461.4 | NP_611305.1 | 5 - 6 | 972 | 67 | |
GenBank mRNA | AY060610.1 | - | 5 - 6 | 939 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |