Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip55E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299649.1 | NP_001286578.1 | 1 | 1136 | 71 | |
NM_137508.4 | NP_611352.1 | 1 | 1136 | 71 | ||
GenBank mRNA | AY089545.1 | - | 1 | 1069 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |