Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EndoB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103905.3 | NP_001097375.1 | 3 - 4 | 728 | 49 | |
NM_137566.4 | NP_611410.1 | 3 - 4 | 728 | 49 | ||
GenBank mRNA | AY071608.1 | - | 3 - 4 | 755 | 49 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |