Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274158.1 | NP_001261087.1 | 10 - 11 | 1672 | 104 | |
NM_137567.3 | NP_611411.2 | 9 - 10 | 1624 | 104 | ||
NM_166341.3 | NP_725875.3 | 16 - 17 | 5121 | 104 | ||
GenBank mRNA | BT010242.1 | - | 9 - 10 | 1618 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |