Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oseg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137582.3 | NP_611426.1 | 3 - 4 | 1611 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |