Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oseg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137582.3 | NP_611426.1 | 7 - 8 | 3739 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |