Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oseg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137582.3 | NP_611426.1 | 8 - 9 | 4108 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |