Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137655.3 | NP_611499.3 | 1 - 2 | 1532 | 68 | |
GenBank mRNA | AY274835.1 | - | 1 - 2 | 1517 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |