Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103917.3 | NP_001097387.1 | 3 - 4 | 955 | 92 | |
NM_001274182.2 | NP_001261111.1 | 3 - 4 | 955 | 92 | ||
NM_137678.3 | NP_611522.1 | 3 - 4 | 955 | 92 | ||
GenBank mRNA | AY061821.1 | - | 3 - 4 | 940 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |