Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137689.5 | NP_611533.2 | 5 - 6 | 1612 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |