Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299732.1 | NP_001286661.1 | 5 - 6 | 3861 | 107 | |
NM_001299733.1 | NP_001286662.1 | 5 - 6 | 3630 | 107 | ||
NM_137707.3 | NP_611551.1 | 5 - 6 | 4158 | 107 | ||
GenBank mRNA | AY060380.1 | - | 5 - 6 | 4129 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |