Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032275.2 | NP_001027446.2 | 5 - 6 | 2194 | 58 | |
NM_001103925.3 | NP_001097395.1 | 5 - 6 | 1561 | 58 | ||
NM_001103926.4 | NP_001097396.3 | 6 - 7 | 2314 | 58 | ||
NM_001299741.1 | NP_001286670.1 | 5 - 6 | 1561 | 58 | ||
GenBank mRNA | KJ826443.1 | - | 4 - 5 | 799 | 58 | |
KJ826444.1 | - | 4 - 5 | 799 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |