Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272589.1 | NP_001259518.1 | 7 - 8 | 2092 | 60 | |
NM_001272591.1 | NP_001259520.1 | 8 - 9 | 2740 | 60 | ||
NM_167374.3 | NP_727717.1 | 7 - 8 | 2092 | 60 | ||
GenBank mRNA | AY119073.1 | - | 6 - 7 | 1130 | 60 | |
AY124548.1 | - | 6 - 7 | 1130 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |