Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298310.1 | NP_001285239.1 | 8 - 9 | 1643 | 59 | |
NM_167408.2 | NP_727781.1 | 9 - 10 | 1764 | 59 | ||
GenBank mRNA | AY060637.1 | - | 9 - 10 | 1764 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |