Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acj6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014743.3 | NP_001014743.1 | 1 - 2 | 872 | 76 | |
NM_001014744.2 | NP_001014744.1 | 1 - 2 | 872 | 76 | ||
NM_001258753.2 | NP_001245682.1 | 1 - 2 | 872 | 76 | ||
NM_001258757.2 | NP_001245686.1 | 1 - 2 | 872 | 76 | ||
NM_080137.3 | NP_524876.1 | 1 - 2 | 872 | 76 | ||
NM_167438.2 | NP_727845.1 | 1 - 2 | 872 | 76 | ||
GenBank mRNA | AF086816.1 | - | 1 - 2 | 262 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |