Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hang | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144732.2 | NP_001138204.1 | 5 - 6 | 4839 | 92 | |
NM_167520.3 | NP_727979.3 | 5 - 6 | 4047 | 92 | ||
NM_167521.3 | NP_727980.2 | 5 - 6 | 4035 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |