Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370033.1 | NP_001356948.1 | 2 - 3 | 1674 | 71 | |
NM_167558.2 | NP_728054.1 | 2 - 3 | 1674 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |