Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298446.1 | NP_001285375.1 | 3 - 4 | 561 | 67 | |
NM_132993.3 | NP_573221.2 | 3 - 4 | 561 | 67 | ||
NM_167569.2 | NP_728082.2 | 2 - 3 | 567 | 67 | ||
NM_167570.2 | NP_728083.2 | 1 - 2 | 260 | 67 | ||
GenBank mRNA | AY122241.1 | - | 2 - 3 | 567 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |