Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201739.2 | NP_001188668.1 | 13 - 14 | 4332 | 74 | |
NM_001258798.2 | NP_001245727.1 | 15 - 16 | 3942 | 74 | ||
NM_001316640.1 | NP_001303569.1 | 12 - 13 | 2585 | 74 | ||
NM_167594.4 | NP_728122.1 | 13 - 14 | 4332 | 74 | ||
NM_167595.4 | NP_728123.1 | 12 - 13 | 2585 | 74 | ||
NM_206775.5 | NP_996498.2 | 15 - 16 | 3939 | 74 | ||
GenBank mRNA | X07132.1 | - | 2213 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster Inventoried |