Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wupA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258803.2 | NP_001245732.1 | 3 - 4 | 333 | 62 | |
NM_167608.3 | NP_728142.1 | 3 - 4 | 333 | 62 | ||
GenBank mRNA | BT125790.1 | - | 3 - 4 | 302 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |