Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298481.1 | NP_001285410.1 | 2 - 3 | 982 | 64 | |
NM_167626.2 | NP_728186.1 | 2 - 3 | 982 | 64 | ||
GenBank mRNA | AB085747.1 | - | 2 - 3 | 869 | 64 | |
AY118385.1 | - | 2 - 3 | 977 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |