Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078682.4 | NP_523406.1 | 3 | 1796 | 81 | |
NM_167633.2 | NP_728201.1 | 1 | 1944 | 81 | ||
NM_167634.3 | NP_728202.1 | 2 | 1981 | 81 | ||
NM_167635.3 | NP_728203.1 | 2 | 1702 | 81 | ||
GenBank mRNA | AY060682.1 | - | 1 | 1961 | 81 | |
X63828.1 | - | 1785 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |