Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133144.4 | NP_573372.1 | 3 - 4 | 1027 | 73 | |
NM_167642.3 | NP_728235.1 | 3 - 4 | 1027 | 73 | ||
GenBank mRNA | AY069536.1 | - | 3 - 4 | 1016 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |