Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103560.3 | NP_001097030.1 | 5 - 6 | 1763 | 57 | |
NM_133144.4 | NP_573372.1 | 4 - 5 | 1139 | 57 | ||
NM_167642.3 | NP_728235.1 | 4 - 5 | 1139 | 57 | ||
GenBank mRNA | AY069536.1 | - | 4 - 5 | 1128 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |